NOTE
The System Check option is not available if the calibration is expired.
The System Check routine performs the following steps:
- Checks the spray stability. If the spray stability check passes, the routine runs the next check.
- Checks the EM Gain calibration.
- Checks the mass position and resolution calibration by using the measured EMRS spectrum.
If any check fails, the System Check routine automatically reruns the calibration process for the failed check.
Prerequisites
- Prepare the MS for calibration. See Prepare the MS system for calibration.
Procedure
- Select the Calibration tab.
- In the Calibration pane, select the System Check option.
- Select Start.
- The Stop button replaces the Start button and the routine begins.
Figure System Check routine in progress - When the calibration is complete, the Tune application prompts you to generate a report of the calibration results.
Figure Report Generation Options - At the prompt, choose an option and select OK.