After you start the flow of the calibration solution to the ion source, monitor the full scan spectrum of the calibration solution.
Procedure
  1. If you have not already applied the favorites setting for the polarity mode you are calibrating, apply it now.
  2. The favorites setting for the positive polarity mode sets the scan mode to Full Scan (Q1) with a mass range from 50 to 1650 m/z.
  3. Inspect the spectrum in the spectrum view of the Tune window.
  4. For the positive mode, check the peaks at the following m/z values:
    69, 102, 215, 622, 922, and 1522.
  5. For the negative mode, check the peaks at the following m/z values:
    69, 113, 302, 602, 1034, and 1634.
  6. For each of the expected peaks, check that the peak is the most abundant peak within ±10 amu of the expected m/z value.
  7. If necessary, adjust the gas settings in the Ion Source pane, especially the Aux Gas parameter setting or the Sweep Gas parameter setting.
  8. Check for the following interfering or missing m/z values:
  9. For the positive mode, check for contaminating peaks at m/z 100 and m/z 212. Possible sources of the m/z 100 contaminant include the nitrogen line or the ion source housing. The contaminant at m/z 212 is probably caused by wearing nitrile gloves while working with the EMRS.
  10. For the negative mode, check for a missing or small peak at m/z 302. The peak at m/z 302 disappears as the EMRS degrades. If the peak is too small or missing, fill the syringe with fresh solution.