CAUTION
This is an emergency shut down procedure. Do not shut down the MS for routine maintenance of the API source interface components.
If you must shut down the MS in an emergency, follow this procedure.
Procedure
  1. To shut down the MS in an emergency, place the Main Power switch in the Off position.
  2. The power switch is located on the power panel on the right-side of the MS.
  3. This switch turns off all power to the MS and any forepumps connected through contact closure, without harming any components within the instrument.
  4. If you cannot safely reach the main power switch, unplug the power cord from the laboratory wall outlet.
  5. Figure Main power switch on the MS power module in the On position
  1. To turn off the LC, autosampler, and data system computer in an emergency, use their respective on/off switches or buttons.