Flush the spray insert after running samples, if the system will be idle afterwards. You can use the syringe pump or an LC pump to flush a 50:50 methanol/water solution through the spray insert at a flow rate of 200 to 400 μL/min for approximately 15 minutes.
NOTE
If you have just run the mass calibration procedure with the EMRS, flush the inlet components with a 50:50 acetonitrile/water solution.
TIP
You do not need to flush the inlet components daily. Flush the inlet components if a mass spectrum shows unwanted contamination peaks.
CAUTION
When the MS’s ion transfer tube is installed, do not flush it with cleaning solution, which flushes the residue into the MS.
Procedure
- Turn off the liquid flow from the syringe pump.
- Place the MS in Standby mode.
- Remove the syringe from the syringe pump as follows:
- Lift the syringe holder off of the syringe.
- Press the pusher block’s release button and slide the block to the left.
- Remove the syringe from the holder.
- Carefully remove the syringe needle from the Teflon™ tube on the syringe adapter assembly.
- Rinse the syringe with a solution of 50:50 methanol/water.
- Flush the sample transfer line, sample tube, and spray insert as follows:
- Load the clean syringe with a solution of 0.1% formic acid in 50:50 methanol/water (or another appropriate solvent, such as a solution of 0.1% formic acid in 50:50 acetonitrile/water).
- Carefully reinsert the syringe needle into the Teflon tube on the syringe adapter assembly.
- Slowly depress the syringe plunger to flush the solution through the sample transfer line, sample tube, and spray insert.
- Remove the syringe needle from the syringe adapter assembly.