You can check the current pressure values in the ion transfer tube-RF lens, and the foreline region (labeled Source Pressure), and in the analyzer region (labeled Analyzer Pressure) in the Status pane of the Tune window.
CAUTION
For proper performance, operate the system at the proper vacuum levels. Poor vacuum levels can cause reduced sensitivity and reduced electron multiplier life.
Procedure
  1. Open the Tune window.
  2. Select the Status tab and select Expand next to Vacuum.
  3. Compare the current values of the pressures in the vacuum manifold with the values listed in the following tables.
  4. After powering up the system, monitor the vacuum readings and confirm they are decreasing with time. Wait until the readings are within typical pressure ranges for your MS. If the pressure remains high, contact your Thermo Fisher Scientific representative for assistance.
  5. If the values are higher than normal, you might have an air leak.
Table Typical pressure readings (TSQ Altis Plus and TSQ Altis)
Conditions
Source pressure (Torr)
(foreline, ion transfer tube, RF lens region)
Analyzer pressure (Torr)
(analyzer region after pumping for at least 12 hours)
Note: The source pressure reading is based on the ion transfer tube set to a temperature of 325 ºC.
Collision gas off, ion transfer tube orifice sealed
<0.5
Less than 6 × 10–6
Collision gas off, ion transfer tube orifice open
<3.5
Less than 9 × 10–6
Collision gas set to 1.5 mTorr, ion transfer tube orifice open
<3.5
Approximately 3 × 10–5
Table Typical pressure readings (TSQ Quantis Plus and TSQ Quantis)
Conditions
Source pressure (Torr)
(foreline, ion transfer tube, RF lens region)
Analyzer pressure (Torr)
(analyzer region after pumping for at least 12 hours)
Note: The source pressure reading is based on the ion transfer tube set to a temperature of 325 ºC.
Collision gas off, ion transfer tube orifice sealed
<0.5
Less than 6 × 10–6
Collision gas off, ion transfer tube orifice open
<2.0
Less than 9 × 10–6
Collision gas set to 1.5 mTorr, ion transfer tube orifice open
<2.0
Approximately 3 × 10–5
Table Typical pressure readings (TSQ Fortis Plus and TSQ Fortis)
Conditions
Source pressure (Torr)
(foreline, ion transfer tube, RF lens region)
Analyzer pressure (Torr)
(analyzer region after pumping for at least 12 hours)
Note: The source pressure reading is based on the ion transfer tube set to a temperature of 300 ºC.
Collision gas off, ion transfer tube orifice sealed
<0.5
Less than 6 × 10–6
Collision gas off, ion transfer tube orifice open
<2.0
Less than 9 × 10–6
Collision gas set to 1.5 mTorr, ion transfer tube orifice open
<2.0
Approximately 3 × 10–5
Table Typical pressure readings (TSQ Quantiva)
Conditions
Source pressure (Torr)
(foreline, ion transfer tube, RF lens region)
Analyzer pressure (Torr)
(analyzer region after pumping for at least 12 hours)
Note: The source pressure reading is based on the ion transfer tube set to a temperature of 350 ºC.
Collision gas off, ion transfer tube orifice sealed
<0.5
Less than 6 × 10–6
Collision gas off, ion transfer tube orifice open
3.8–4.2
Less than 9 × 10–6
Collision gas set to 1.5 mTorr, ion transfer tube orifice open
3.8–4.2
Approximately 3 × 10–5
Table Typical pressure readings (TSQ Endura)
Conditions
Source pressure (Torr)
(foreline, ion transfer tube, RF lens region)
Analyzer pressure (Torr)
(analyzer region after pumping for at least 12 hours)
Note: The source pressure reading is based on the ion transfer tube set to a temperature of 350 ºC.
Collision gas off, ion transfer tube orifice sealed
<0.5
Less than 6 × 10–6
Collision gas off, ion transfer tube orifice open
<2.0
Less than 9 × 10–6
Collision gas set to 1.5 mTorr, ion transfer tube orifice open
<2.0
Approximately 3 × 10–5