Check both argon and nitrogen regulators to determine how much gas is in the tank.
Gas | Procedure |
---|---|
Argon | Procedure
|
Nitrogen | Procedure
|
CAUTION
Do not operate the MS without nitrogen flow as this can damage the source and front end optics due to particulate matter in the atmosphere. Before you operate the MS, verify that there is sufficient nitrogen for the API source.
If the system runs out of nitrogen while you are acquiring scans in the Tune application, a prompt appears with a warning that the nitrogen supply is probably empty. If you are running an acquisition sequence, the data system completes the current run. The system does not proceed to the next sample because the MS cannot return to the Ready state when the nitrogen set point cannot be reached. Meanwhile, the MS continues scanning.
For more information about gas requirements, refer to the Preinstallation Requirements Guide for your MS.