To reduce downtime, these topics describe the tasks that you can do before and after you operate the mass spectrometer:
NOTE
You do not need to calibrate and tune the MS as part of your daily routine. Generally, you must calibrate the MS every one to three months of operation for optimum performance over the entire mass range of the mass detector.