The three rod assemblies are numbered from the API source end of the manifold and are designated Q1, Q2, and Q3. Quadrupoles Q1 and Q3 enable high-resolution scans without signal loss.
For details, see these topics:
The three rod assemblies are numbered from the API source end of the manifold and are designated Q1, Q2, and Q3. Quadrupoles Q1 and Q3 enable high-resolution scans without signal loss.
For details, see these topics: