The TSQ Fortis Plus and TSQ Fortis instruments have a Matrix Separator Ion Guide (MSIG), which consists a tube lens and skimmer. Ions from the ion transfer tube pass through the tubes lens and then the skimmer.
Figure Tube lens (left) and skimmer (right)
Tube lens (left) and skimmer (right)