When you remove the ion transfer tube (after it has cooled to room temperature), the vent prevent ball of the API source interface drops into place to stop air from entering the vacuum manifold. By acting as a barrier, the vent prevent ball lets you remove the ion transfer tube for cleaning or replacement without venting the system.
Figure Vent prevent ball positions
12
No.DescriptionNo.Description
1
Ion transfer tube installed with vent prevent ball elevated
2
Ion transfer tube removed with vent prevent ball dropped
For information about cleaning or replacing the ion transfer tube, see Maintain the external components of the API source interface.