The heater block of the API source interface contains two heater cartridges that surround the ion transfer tube and heat the tube to temperatures up to 400 °C (752 °F). A decreasing pressure gradient draws ions into the ion transfer tube in the atmospheric pressure region.