The spray cone is an integral component of the API source interface cage. The ion transfer tube fits through a small orifice in the center of the spray cone. A graphitized Vespel™ seal acts as a seal between the spray cone and the back of the ion transfer tube’s nose cone.
The custom tool that you use to remove and install the ion transfer tube depends on whether the ion transfer tube is secured to the spray cone with a quarter-turn pin or a threaded fitting.
The following TSQ Series II mass spectrometers use an ion transfer tube that is secured with a quarter-turn pin:
  • TSQ Altis Plus
  • TSQ Altis
  • TSQ Quantiva
Spray cone (with a quarter-turn bushing) with the ion transfer tube and graphitized Vespel O-ring removed
1234
No.DescriptionNo.Description
1
High-capacity ion transfer tube for the TSQ Altis Plus and the TSQ Altis
3
Bushing for the quarter-turn ion transfer tube
2
Graphitized Vespel seal
4
Spray cone
NOTE
The ion transfer tube for the TSQ Quantiva MS is also a high-capacity transfer tube, but its nose cone does not have any etchings. See Ion transfer tube.
Spray cone with the ion transfer tube and O-ring removed (TSQ Quantis Plus and TSQ Quantis)
123
No.DescriptionNo.Description
1
Ion transfer tube for the TSQ Quantis Plus MS and the TSQ Quantis MS
3
Spray cone with a threaded entry port
2
Graphitized Vespel seal
NOTE
The ion transfer tubes for the TSQ Fortis Plus, TSQ Fortis, and TSQ Endura instruments are also secured to the spray cone with a threaded fitting.