The following topics describe the components of the API source interface:
Except for the atmospheric pressure side of the spray cone and the nose cone of the ion transfer tube that is within the API source, the API source interface components are located in a vacuum chamber.
The following figure shows a cross-section schematic of the API source interface for the TSQ Altis Plus and TSQ Altis instruments. The API source interface for the TSQ Quantiva MS is similar to this source.
Figure API source interface cross section (TSQ Altis Plus and TSQ Altis)
123456789
No.DescriptionNo.Description
1
Ion sweep cone
6
L0 lens
2
Vent prevent ball
7
MP00 multipole
3
Ion transfer tube
8
Heater block
4
Electrodynamic Ion Funnel (EDIF)
9
Spray cone
5
Exit lens
The following figure shows a cross-section schematic of the API source interface for the TSQ Fortis Plus and the TSQ Fortis instruments.
Figure API source interface cross section (TSQ Fortis Plus and TSQ Fortis)
123456789
No.DescriptionNo.Description
1
Ion sweep cone
6
L0 lens
2
Vent prevent ball
7
MP00 multipole
3
Ion transfer tube
8
Heater block
4
Tube lens
9
Spray cone
5
Skimmer
The following figure shows a cross-section schematic of the API source interface for the TSQ Endura MS. The API source interface for the TSQ Quantis Plus and TSQ Quantis instruments is similar to this source.
Figure API source interface cross section (TSQ Endura)
123456789
No.DescriptionNo.Description
1
Ion sweep cone
6
L0 lens
2
Vent prevent ball
7
MP00 multipole
3
Ion transfer tube
8
Heater block
4
Stacked Ring Ion Guide (SRIG)
9
Spray cone
5
Exit lens