You can apply nitrogen gas at the API source by using any combination of the three gas sources: Sheath gas, Auxiliary gas, and (optional) Sweep gas.
Sheath gas is the inner-coaxial nitrogen gas that helps nebulize the sample solution into a fine mist as the solution exits the API spray insert nozzle.
Sheath gas flow for ESI source (blue lines)
12
No.DescriptionNo.Description
1
Sheath gas
2
Electrospray needle
Auxiliary gas is the outer-coaxial nitrogen gas that helps the sheath gas to nebulize and evaporate the sample solution by focusing the vapor plume and lowering the humidity in the API source.
Auxiliary gas flow (green lines)
12
No.DescriptionNo.Description
1
Auxiliary gas
2
Electrospray needle
Sweep gas is the off-axis nitrogen gas that flows out from behind the optional ion sweep cone to aid in solvent declustering and adduct reduction. The optional ion sweep cone has an inlet for the sweep gas.
All analyses are analyte dependent and require separate optimization with sheath gas, sweep gas, and auxiliary gas to determine which combination yields optimum performance. Optimize with each gas independently before you perform experiments using MSn techniques and before you perform any quantitative analysis experiments.
For higher LC flow rates, apply auxiliary gas to increase the signal intensity.
When you are analyzing complex matrices such as plasma or nonvolatile salt buffers, you must use sweep gas to increase the ruggedness of the system. For full-scan MS or data-dependent scan experiments, apply sweep gas to improve the signal-to-noise ratio.