The following figure shows the vacuum manifold with the top cover plates removed next to the turbomolecular pump. The vacuum manifold is a thick-walled, aluminum chamber that encloses the API source interface, ion optics, mass analyzer, and ion detection system assemblies. It has multiple removable top cover plates, various electrical feedthroughs, and gas inlets.
Placement of the turbomolecular pump next to the vacuum manifold
123456789
No.DescriptionNo.Description
1
API source interface
6
Q2 collision cell
2
RF lens and ion transfer tube chamber
7
Q3 mass analyzer chamber
3
MP00 ion optics chamber
8
Ion detector
4
MP0 ion optics chamber (also known as Q0)
9
Turbomolecular pump
5
Q1 mass analyzer chamber
The following table lists the typical pressures in region 2 (which is evacuated by the forepump or forepumps) and region 5 (which is evacuated by the triple-inlet turbomolecular pump).
Typical pressures in regions 2 and 5
Model
Region 2
Source pressure readback
Region 5
Analyzer pressure readback
TSQ Altis Plus
3–4 Torr
3–7 × 10–6 Torr
TSQ Quantis Plus
1–2 Torr
2–6 × 10–6 Torr
TSQ Fortis Plus
1–2 Torr
1–6 × 10–6 Torr
TSQ Altis
3–4 Torr
3–7 × 10–6 Torr
TSQ Quantis
1–2 Torr
2–6 × 10–6 Torr
TSQ Fortis
1–2 Torr
1–6 × 10–6 Torr
TSQ Quantiva
2–6 × 10–6 Torr
3–7 × 10–6 Torr
TSQ Endura
3–7 × 10–6 Torr
2–6 × 10–6 Torr