The FAIMS Pro and FAIMS Pro Duo interfaces are compatible with all TSQ Series II mass spectrometers (GLE models) except for the TSQ Fortis and TSQ Fortis Plus instruments.
The FAIMS Pro and FAIMS Pro Duo interfaces are compatible with all TSQ Series II mass spectrometers (GLE models) except for the TSQ Fortis and TSQ Fortis Plus instruments.