The following table lists the ion sources that are compatible with the TSQ Series II mass spectrometers.
Hardware | Flow rate range | Compatible models |
|---|---|---|
OptaMax NG API source | H-ESI: 1–1,000 μL/min APCI: 100–2,000 μL/min | TSQ Altis Plus |
Ion Max NG API source | H-ESI: 1–1,000 μL/min APCI: 100–2,000 μL/min | TSQ Quantiva |
EASY-Max NG API source | H-ESI: 1–1,000 μL/min APCI: 100–2,000 μL/min | TSQ Endura |
EASY Spray NG | 10–1,000 nL/min | All GLE models |
OptaMax Duet | H-ESI: 1–1000 μL/min APCI: 100–2,000 μL/min | All GLE models |
VeriSpray PaperSpray | N/A | All GLE models |