The following table lists the specifications for the models in the TSQ Series II product line.
MS model | Relative cost | Mass range | Scan speed | Selectivity |
|---|---|---|---|---|
Third generation | ||||
TSQ Altis Plus | High | 2–2010 | 800 | 0.2 Da FWHM |
TSQ Quantis Plus | Mid-range | 2–3000 | 800 | 0.4 Da FWHM |
TSQ Fortis Plus | Low | 2–3000 | 800 | 0.4 Da FWHM1) |
Second generation | ||||
TSQ Altis | High | 2–2000 | 600 | 0.2 Da FWHM |
TSQ Quantis | Mid-range | 2–3000 | 600 | 0.4 Da FWHM |
TSQ Fortis | Low | 2–3000 | 600 | 0.4 Da FWHM |
First generation | ||||
TSQ Quantiva | High | 2–1800 | 500 | 0.2 Da FWHM |
TSQ Endura | Low | 2–3400 | 500 | 0.4 Da FWHM |