The basic components of a mass spectrometer are the ion source, the mass analyzer, and the ion detector. The following figure shows a schematic of the TSQ Altis Plus MS.
Figure TSQ Altis Plus MS schematic
123456789
No.
Description
No.
Description
1
Q2 mass analyzer or active collision cell with axial DC field
(Q2 quadrupole can behave as an ion transmission device or a collision cell with axial DC field)
6
RF lens, electrodynamic ion funnel (EDIF) for the TSQ Altis Plus MS and TSQ Altis MS
2
Q3 mass analyzer (segmented quadrupoles with a hyperbolic surface)
7
MP00 assembly (MP00 cage, MP00 multipole, exit lens, and L0 lens)
3
Dual-mode electron multiplier detector
8
Ion beam guide with neutrals blocker
4
Sweep cone
9
Q1 mass analyzer (segmented quadrupoles with a hyperbolic surface)
5
Ion transfer tube