The drainage system for a TSQ Series II MS begins with the drainage port at the bottom of the API source interface on front panel of the MS and ends with the waste container that connects to the drain port on the left side of the MS. An internal tubing connects the drainage port at the bottom of the API source interface to the drain port on the left side of the MS.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Drainage port under the API source interface | 4 | Waste container |
2 | Internal drainage tubing | 5 | Connection to a dedicated lab exhaust system |
3 | Drain/waste port on the left side of the MS |