The MS receives line power at 230 Vac ±10%, 5 A, 50/60 Hz through the power entry module located on the right-side panel of the MS.
Figure Power entry module
12345
No.DescriptionNo.Description
1
Electronics service switch
4
Power In receptacle (230 Vac)
2
Pump enable connector (mechanical forepump on/off control)
5
Main Power switch
3
AC Output receptacle (reserved for future use)
Main power switch
In the Off position, the Main Power (circuit breaker) switch removes all power to the MS. In the On position, the MS receives power. In the standard operational mode, the switch stays in the On position.
Placing the Main Power switch in the Off position turns off any forepump connected to the MS through contact closure, but it does not turn off line power to the forepump, as it connects directly to the laboratory line power.
CAUTION
In an emergency, do not use the Electronics service switch to shut off power to the MS. Instead, place the main power circuit breaker switch (labeled Main Power) in the Off (down) position and disconnect the power cord from the electrical outlet.
Electronics service switch
In the Service Mode (down) position, the switch removes power to all components of the MS except for the fans and vacuum system. This setting allows you to service non-vacuum system components with the vacuum system still operating. In the Operating Mode (up) position, all components of the MS have power.
Pump enable connector (mechanical forepump)
The MS starts and stops the required forepump by using a relay control cable that connects to the forepump’s contact closure connector.