The following table describes the LED indicators on the front panel of the MS.
LED | State | Description |
|---|---|---|
Power | Green |
|
Off |
| |
Vacuum | Green | The vacuum is within the allowable operating range. |
Yellow | The vacuum is outside the allowable operating range. | |
Off | The MS is either off or in the process of starting up. | |
Communication | Green | The MS and data system are communicating. |
Yellow | The MS and data system are trying to establish a communication link. | |
Off | The MS is off. | |
System | Green | The MS is on. |
Yellow | The MS is in Standby mode. | |
Off | The MS is off. | |
Scan | Flashing blue | The MS is on and scanning. |
Off | The MS is not scanning. |