These are the software installation instructions for the Thermo Scientific™ TSQ Series II 4.01) instrument control software.
NOTE
You must have local administrator privileges on the data system computer to install the Thermo Scientific software.
NOTE
The TSQ Series II 4.0 instrument control software is compatible with the following software packages that support various liquid chromatography (LC) instruments:
  • Chromeleon CDS (chromatography data system) 7.3.2 MUc or later
  • Chromeleon CDS (chromatography data system) 7.2.10 MUh or later
  • SII (standard instrument integration) for Xcalibur 1.7 with hotfix or later
  • Aria MX 2.7 SP1 or later
  • LC Devices 3.2 SP3 or later
Depending on your system requirements, perform one of the following procedures: