The Status pane displays real-time readback information for various components in the MS. The color-coded status indicators update every few seconds and are as follows:
  • Green: Indicates that the parameter is within its expected range.
  • Orange: Indicates that the parameter is trending toward the expected range.
  • Red: Indicates that the parameter is outside of its expected range.
Figure Status pane
123
No.DescriptionNo.Description
1
Colored status readback indicators
3
Status pane button
2
By Board or By Function selection
Procedure
  1. Select the Status pane.
  2. From the drop-down list in the title bar, do one of the following:
  3. - Select By Board to display a list of components based on the electronic PCB board that controls their operation.
  4. - Select By Function to display a list of components based on their functionality in the MS.
TIP
Select the arrow on the right side of the window to show or hide the Status pane.