The following table describes the parameters for the Full Scan type (Q1 and Q3).
Parameter | Description |
|---|---|
Scan Type | The full scan type performs a single stage of mass analysis, producing a mass spectrum similar to that of a single quadrupole instrument. Select Full Scan (Q1) or Full Scan (Q3) from the drop-down menu. |
Scan Range (m/z) | Specify the scan m/z range. Range: 2–2,010 |
Q1 Resolution (FWHM) | Select the Q1 Resolution (FWHM) options from the following options: Range: 0.2, 0.4, 0.7, 1.2, and 2 |
Q3 Resolution (FWHM) | Select the Q3 Resolution (FWHM) options from the following options: Range: 0.2, 0.4, 0.7, 1.2, and 2 |
Scan Rate (Da/sec) | Select a scan rate from the following options: Range: 250, 500, and 1,000 (default) NOTE: When operating in Full Scan mode using Q3 and a FWHM of 2, additional scan speeds of 5,000 and 20,000 are available. |
CID Gas (mTorr) | Select a CID gas value from the following options: Range: 0, 0.5, 1, 1.5, 2, 2.5, 3, 3.5, and 4 |
Source Fragmentation | Specify the collision energy (in volts) for ion source fragmentation. Range: 0–140 |