The following figure shows an example of the Alerts Console when minimized at the bottom of the Tune window.
Figure Alerts Console minimized
12345
No.DescriptionNo.Description
1
Error alert
4
Information alert
2
Warning alert
5
Alerts Console minimized at bottom of Tune window
3
Recovered errors and warnings alert
To expand the minimized Alerts Console, select an area near the count icons. This action opens a docked window that displays the alert details, as shown in the following figure.
Figure Alerts Console displayed as a docked window
123
No.DescriptionNo.Description
1
Event table
3
Maximize button
2
Docked window
To switch from the docked window to a floating window, select the Maximize button located on the right side. In a floating window, the Alerts Console displays the full event table along with additional control options. The event table can display up to 1,000 alerts. Error and Warning alerts remain visible until they are recovered, while Informational alerts are removed once the 1,000-entry limit is reached. You can organize and sort events by selecting the column headers in the table.
The following figure shows the Alerts Console as a floating window.
Figure Alerts Console displayed as a floating window
12
No.DescriptionNo.Description
1
Control options
2
Event table