The following table describes the parameters in the Directed DDA Scan Table.
NOTE
Some parameters in the Properties pane can be added to the table. Select the table icon next to the parameter, then enter the appropriate value in the column.
Parameter
Description
Scan Range (m/z)
Specify the scan range.
Range: 2–2,010
NOTE: The Scan Range and Scan Rate are interdependent.
Scan Rate (Da/sec)
The rate at which the quadrupole scan across the mass range.
Range: 250, 500, and 1,000; default: 1,000
Q1 Resolution (FWHM)
FWHM resolution for ions passed by the Q1 mass analyzer. As the FWHM value is decreased, resolution is increased giving a greater selectivity.
Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7
Intensity Threshold
When an ion exceeds the specified threshold, a DDA scan is triggered.
Range: 0–1e9; default: 0.0e0