The following table describes the parameters in both the SIM Q1 Properties and SIM Q3 Properties.
Parameter
Description
Polarity
By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon to remove the column from the table. Then select the ionization mode (positive or negative).
Data Mode
Select the data mode (Profile or Centroid).
Scan Option
Select the scan type option:
  • Scan Rate
  • Dwell Time
Scan Rate (Da/sec)
(Available when the Scan Option is set to Scan Rate.)
The rate at which the quadrupole scan across the mass range.
Range: 250, 500, and 1,000; default: 250
NOTE: In a SIM Q3 Scan, when the Q3 resolution (FWHM) is set to 2, the 5,000 and 20,000 options become available.
Scan Width (m/z)
(Available when the Scan Option is set to Scan Rate.)
Specifies the scan width around the Center Mass. For example, a scan width of 10 around a center mass of 200 scans from m/z 195 to 205.
Range: 0.5–50; default: 10
Dwell Time (ms)
(Available when the Scan Option is set to Dwell Time.)
Specifies the time the instrument scans each transition. The dwell time is the period the mass analyzer remains on a peak. Increasing the dwell time improves ion statistics but also increases the overall cycle time.
Range: 0.5–1,000; default: 40
Use Calibrated RF Lens
Uses the mass-dependent RF lens voltages from the calibration lookup table instead of a compound-dependent value.
Default: Selected
Q1 Resolution (FWHM)
(Available in the SIM Q1 scan type.)
FWHM resolution for ions passed by the Q1 mass analyzer. As FWHM value decreased, resolution is increased giving a greater selectivity.
Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7
Q3 Resolution (FWHM)
(Available in the SIM Q3 scan type.)
FWHM resolution for ions passed by the Q3 mass analyzer.
Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7
Source Fragmentation
Voltage applied to induce CID ion source fragmentation.
Range: 0–140
Chromatographic Peak Width (sec)
Average width at the base of the chromatographic peak. Ignore this field when using infusion.
The MS measures the chromatographic peak width at the base of the peak and applies a low-pass filter to remove high-frequency noise from the signal.
Range: 0–100; default: 6
NOTE: This option is unavailable when Mixed Scan Mode is selected.
Use Chromatographic Filter
Uses the chromatographic high frequency noise filter, which is calculated from the chromatographic peak width.
Default: Selected
Display Retention Time
Deselect to view retention time as Start Time and End Time instead of Retention Time and RT Window in the SRM table.
Default: Selected