NOTE
Some parameters in the Properties pane can be added to the table. Select the table icon next to the parameter, then enter the appropriate value in the column.
The following table describes the parameters in the Neutral Loss Scan Table.
Parameter
Description
Neutral Loss (m/z)
Precursor ions fragment into both charged and neutral fragments. Enter the common neutral loss for a group of compounds.
Range: 2–2,010; default: 98
Polarity
Select the ionization mode (positive or negative). Both positive and negative scan events can be used in the same method.
NOTE: By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon to remove the column from the table.
Scan Range (m/z)
Specify the scan range.
Range: 2–2,010
NOTE: The Scan Range and Scan Rate are interdependent.