The following table describes the parameters in the Product Ion Scan Properties pane.
Parameter | Description |
|---|---|
Polarity | By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon |
Data Mode | Select the data mode (Profile or Centroid). |
Scan Rate (Da/sec) | The rate at which the quadrupole scan across the mass range. Range: 250, 500, and 1,000; default: 1,000 NOTE: In a Full Scan Q3, when the Q3 resolution (FWHM) is set to 2, the 5,000 and 20,000 options become available. |
Use Collision Energy Ramp (CER) | The collision energy is ramped from the Start (CER) when scanning low m/z fragments to the End (CER) at high m/z fragments. If Start (CER) is higher than the End (CER), this is termed a reversed energy ramp and will give a richer product ion spectra, often including the precursor m/z. Default: Deselected |
CER Start (V) | (Available when the Use Collision Energy Ramp (CER) checkbox is selected.) Collision energy at the low m/z of the product ion scan range. Range: 0–90; default: 55 |
CER End (V) | (Available when the Use Collision Energy Ramp (CER) checkbox is selected.) Collision energy at the high m/z of the product ion scan range. Range: 0–90; default: 20 |
Collision Energy (V) | RF amplitude (in volts) used to fragment ions. Range: 0–90; default: 0 |
Use Calibrated RF Lens | Uses the mass-dependent RF lens voltages from the calibration lookup table instead of a compound-dependent value. Default: Selected |
Q1 Resolution (FWHM) | FWHM resolution for ions passed by the Q1 mass analyzer. As the FWHM value is decreased, resolution is increased giving a greater selectivity. Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7 |
Q3 Resolution (FWHM) | FWHM resolution for ions passed by the Q3 mass analyzer. Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7 |
CID Gas (mTorr) | CID gas pressure for the Q2 collision cell. Range: 0, 0.5, 1, 1.5, 2, 2.5, 3, 3.5, and 4 |
Source Fragmentation | Voltage applied to induce CID ion source fragmentation. Range: 0–140 |
Chromatographic Peak Width (sec) | Average width at the base of the chromatographic peak. Ignore this field when using infusion. The MS measures the chromatographic peak width at the base of the peak and applies a low-pass filter to remove high-frequency noise from the signal. Range: 0–100; default: 6 NOTE: This option is unavailable when Mixed Scan Mode is selected. |
Use Chromatographic Filter | Uses the chromatographic high frequency noise filter, which is calculated from the chromatographic peak width. Default: Selected |