The following table describes the parameters in both the Full Scan Q1 Table and Full Scan Q3 Table.
Parameter
Description
Scan Range (m/z)
Specify the scan range.
Range: 2–2,010
NOTE: The Scan Range and Scan Rate are interdependent.
Scan Rate (Da/sec)
The rate at which the quadrupole scan across the mass range.
Range: 250, 500, and 1,000; default: 1,000
NOTE: In a Full Scan Q3, when the Q3 resolution (FWHM) is set to 2, the 5,000 and 20,000 options become available.
Polarity
Select the ionization mode (positive or negative). Both positive and negative scan events can be used in the same method.
NOTE: By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon to remove the column from the table.
Q1 Resolution (FWHM)
(Available in the Full Scan Q1 scan type.)
FWHM resolution for ions passed by the Q1 mass analyzer. As FWHM value decreased, resolution is increased giving a greater selectivity.
Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7
Q3 Resolution (FWHM)
(Available in the Full Scan Q3 scan type.)
FWHM resolution for ions passed by the Q3 mass analyzer.
Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7
Source Fragmentation
Voltage applied to induce CID ion source fragmentation.
Range: 0–140