The following table describes the parameters in the SRM table.
NOTE
Some parameters in the Properties pane can be added to the table. Select the table icon next to the parameter, then enter the appropriate value in the column.
Parameter | Description |
|---|---|
Compound | Specify the name of the chemical compound. The compound name allows grouping of transitions, even if Precursor m/z is different (e.g. +2 and +1 charge states, or Chlorinated isotopes). Compound names are used when viewing data files with the FreeStyle™ software. |
Retention Time (min) | (Available when the Display Retention Time checkbox in the Properties pane is selected.) Specify the mid-point of the retention time window (in minutes). Range: 0–999; default: 1 |
RT Window (min) | (Available when the Display Retention Time checkbox in the Properties pane is selected.) Specify the width of the retention time window (in minutes). The MS monitors transitions only within the user specified time window, which is centered around the retention time. Range: 0–999; default: 1 |
Start Time (min) | (Available when the Display Retention Time checkbox in the Properties pane is deselected.) Time at which acquisition of scan will start. Range: 0–999; default: 0.5 |
End Time (min) | (Available when the Display Retention Time checkbox in the Properties pane is deselected.) Time at which acquisition of scan will end. Range: 0–999; default: 1.5 |
Polarity | Select the ionization mode (positive or negative). Both positive and negative scan events can be used in the same method. NOTE: By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon |
Precursor (m/z) | The precursor mass-to-charge ratio (m/z) transmitted through Q1. Range: 2–2,010 |
Product (m/z) | Specify the mass-to-charge ratio (m/z) for the product ions. Range: 2–2,010 |
Collision Energy (V) | RF amplitude (in volts) used to fragment ions. Range: 0–90; default: 0 |
Reference Threshold | (Available when the Use Retention Time Reference checkbox in the Properties pane is selected.) Retention time reference peak height threshold for the specific SRM transition. Range: 0 to 1 x 109 |
Min Dwell Time (ms) | The minimum dwell time for a transition is calculated based on the number of concurrent SRMs, Cycle Time, and taking into account any interscan delay. |
Dwell Time Priority | (Available when the Use Dwell Time Factor checkbox in the Properties pane is selected.) Select the priority level per analyte:
|