The following table describes the parameters in the SRM Properties pane.
Parameter | Description |
|---|---|
Polarity | By default, the polarity of each scan is defined in the table. To define the polarity globally, select the table icon |
Data Mode | Select the data mode (Profile or Centroid). |
Chromatographic Peak Width (sec) | Average width at the base of the chromatographic peak. Ignore this field when using infusion. The MS measures the chromatographic peak width at the base of the peak and applies a low-pass filter to remove high-frequency noise from the signal. Range: 0–100; default: 6 NOTE: This option is unavailable when Mixed Scan Mode is selected. |
Use Chromatographic Filter | Uses the chromatographic high frequency noise filter, which is calculated from the chromatographic peak width. Default: Selected |
Use Cycle Time | Uses cycle time instead of dwell time. Default: Selected NOTE: This option is unavailable when Mixed Scan Mode is selected. |
Cycle Time (sec) | (Available when Use Cycle Time is selected.) Cycle time is the time desired to analyze all required SRMs at any given time point. Range: 0.002–600; default: 0.6 NOTE: Dwell time for each transition is calculated dynamically. It ensures that scan rate remains constant across the chromatographic peak. Cycle time should be set as: peak width at base / required number of scans. |
Points Per Peak | (Available when Use Cycle Time is selected.) Number of desired data points across the chromatographic peak. Range: 0–5,000 Optimum Range: 5–20; default: 10 NOTE: If you specify a value outside the optimum range (5 to 20), the text box turns yellow. If you specify a value outside the maximum range (0 to 5,000), the text box turns red. |
Use Dwell Time Factor | (Available when Use Cycle Time is selected.) Select to enable dwell time factor. Adds the Dwell Time Priority column to the SRM table. Default: Deselected |
Minimum Preferred Dwell Time (ms) | (Available when Use Dwell Time Factor is selected.) Set preferred minimum dwell time when factored dwell time is used. Dwell time values outside the preferred range will be marked yellow. Range: 0.25–1,000; default: 1 |
Use Calibrated RF Lens | Uses the mass-dependent RF lens voltages from the calibration lookup table instead of a compound-dependent value. Default: Selected |
Q1 Resolution (FWHM) | FWHM resolution for ions passed by the Q1 mass analyzer. As the FWHM value is decreased, resolution is increased giving a greater selectivity. Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 0.7 |
Q3 Resolution (FWHM) | FWHM resolution for ions passed by the Q3 mass analyzer. Range: 0.2, 0.4, 0.7, 1.2, and 2; default: 1.2 |
CID Gas (mTorr) | CID gas pressure for the Q2 collision cell. Range: 0, 0.5, 1, 1.5, 2, 2.5, 3, 3.5, and 4 |
Source Fragmentation | Voltage applied to induce CID ion source fragmentation. Range: 0–140 |
Use Retention Time Reference | Adds the Reference Threshold column to the SRM table to enable Dynamic Retention Time Scheduling. Default: Deselected NOTE: This option is unavailable when Mixed Scan Mode is selected. |
Display Retention Time | Deselect to view retention time as Start Time and End Time instead of Retention Time and RT Window in the SRM table. Default: Selected |
Use Quan Ion | Adds the Use Quan Ion column to the SRM Table. This allows the user to specify which product ion is to be defined as the quan ion when importing a method into the TraceFinder software. Default: Deselected NOTE: You must select at least one row (transition). |
Show Visualization | Select to graphically display the following graphs:
When you update the SRM table, the Method Editor updates the graphs. The graph axes reset if you clear the Show Visualization checkbox. To select or reset a graph, click it. To zoom in, drag the cursor over an area. Default: Deselected NOTE: This option is unavailable when Mixed Scan Mode is selected. |
Copy Experiment Time (button) | Changes either Retention Time and RT Window or Start Time and End Time to span the entire method duration. Converts a scheduled RT method to an unscheduled method. |