Use the SIM Table and the SIM Scan Properties pane to set up a selected ion monitoring (SIM) scan type experiment.
NOTE
  • Hover the pointer over a parameter name to view the tooltip description.
  • When you change a parameter value, it appears in bold until you save the method.
    – A red highlighted parameter box indicates that the value is outside the operational range.
    – A yellow highlighted parameter box indicates that the value is outside the optimum range.
  • Some parameters have a table icon on the left. When selected, the parameter appears as a column in the table, where you can define its value.
  • Select the Summary tab to view the instrument method settings.
For additional information, see the following topics: