The Method Timeline and Settings areas are displayed at the top of both the Global Parameters and Scan Parameters pages. Use the Method Timeline area to create a timeline for your experiment. Each timeline provides a graphical representation of the experiment’s defined time segments.
The following table describes the parameters available in the Method Timeline and Settings area.
Parameter | Description |
|---|---|
Method Timeline | |
Method Duration (min) | Specify the method duration (in minutes). The Method Timeline area displays the remaining experiment time. Range: 0.1–5,000; default: 60 |
Experiment ACTIONS | You can add or delete an experiment in the Method Timeline using the options in the dropdown list:
NOTE: The Add New Experiment option is not available if the Mixed Scan Mode checkbox is deselected. |
Zoom control buttons | These buttons control the timeline display (from top to bottom).
|
| |
Settings | |
Mixed Scan Mode | Select this checkbox to enable multiple experiments in the Method Timeline or multiple scan types to the method on the Scan Parameters page. To create a mixed-scan method, you must select the Mixed Scan Mode checkbox before adding any experiments to the method. If you add a scan mode to a method first, the Mixed Scan Mode checkbox becomes unavailable. Default: Deselected NOTE: If selected, only specific scan types are available in mixed-scan mode because some scan functions are incompatible. |