Procedure
- Select the Scan Parameters tab.
- Select the experiment type that you want to perform from the Quan view, Hybrid view, or Templates view. Then double-click the icon or drag it to the "Place Scan Here" node in the Experiment window.
- Enter your experiment parameters, as applicable.
- Select the Templates View icon and select Save as Template.
- Browse to a destination path, type a file name, and select Save.
- In the Save Method Template dialog box, select one of the following options:
- Current Experiment Only: If there are two or more experiments in the method timeline, only the marked experiment is saved as a method template.
- All Experiments: If there are two or more experiments in the method timeline, all experiments in the method timeline are saved as a method template.
- (Optional) Include Current Global Parameters in the Template: If unchecked, only the scan parameters of the method are saved. If checked, scan parameters and global parameters (such as ion source settings) are saved in the template.
- Select OK to save the template.
- Once saved, the new template is listed under the Templates View > Custom Templates area.
NOTE
Templates must be saved within the Templates directory under a subfolder. You can use the default "My Experiments" folder or you can create custom subfolder.