A report can be generated after running the procedures on the Calibration and Diagnostic panes. Reports can be automatically generated, manually generated, or not generated based on your Tune preferences selection. You can also customize the information that appears in your report.
For details on the various report options, see Tune Preferences.
Prerequisites
- Select the Options
icon and choose Preferences to open the Tune Preferences window.
- Under Report Options, select Show Report Generation Options dialog box.
Procedure
- Perform a calibration or diagnostics routine, as applicable.
- The Report Generation Options dialog box appears.
- Select Generate report(s) and then select OK.
- Save the report as a PDF.