The following table describes the shortcut menu commands in the Spectrum view (by right-clicking).
Parameter | Description |
---|---|
Reset Scaling | Resets the axes to the default intensity and mass ranges. |
Copy to Clipboard | Copies a screen capture of the mass spectrum to the clipboard. |
Pan | To use this tool, you must first zoom in on the spectrum. Then, you can select this tool to slide the spectrum back and forth along the x-axis by holding down the left mouse button and moving the mouse cursor. |
Sends a copy of the mass spectrum to the printer. | |
Show Absolutes | Changes the y-axis scale from percentages to ion flux (ions/s). |
Display |
|
The following table defines the header information at the top of the real-time spectrum.
Header information | Description |
---|---|
# (scan number) | The scan count, which only resets to zero when you start data acquisition. |
RT | The retention time (in minutes). |
NL | The normalization level, which is a measure of the ion signal strength. |
Scan setting | The detector type, polarity mode (+ or –), data type (profile or centroid), source type, scan type, and other settings. |