The following table describes the parameters in the Plot Chromatogram dialog box.
Parameter | Description |
---|---|
Spray Stability | Runs the spray stability evaluation. The spray stability evaluation generates real-time graphs of the TIC, base peak, or user-defined m/z, including the relative standard deviation (RSD). The %RSD must be less than 15 percent for the spray stability evaluation to pass. Default: Cleared |
TIC | Plots the TIC (total ion current) chromatogram. |
Base Peak | Plots the base peak chromatogram. |
User Defined m/z | Plots the signal intensities of the ions you enter in the mass table as a function of time. |
Mass Table | |
Mass | Available when the User Defined m/z option is selected. Indicates up to 10 m/z values for the ion signal intensities that you want to plot. Range: 20–2,000; default: 200 |