The following table describes the buttons and icons available at the top of the Tune application window.
Button | Description |
---|---|
On mode: Indicates the MS is on and is ready for operation. All voltages, the sheath gas, the auxiliary gas, and the sweep gas are on. For additional details on the MS operation, see System Startup, Shutdown, and Reset. NOTE You set the instrument power mode options from the Tune application or data system. When a power mode is active, the center of the selected icon changes from white to green. | |
Standby mode: Indicates the MS is in standby. In standby, the electron multiplier, conversion dynodes, 8 kV power to the source housing, main RF voltage, and ion optic RF voltages are off. The sheath, auxiliary, and sweep gases remain on and return to their default settings (2 arbitrary units). Standby mode is the normal condition to leave the MS in when it is not in use for short periods of time. | |
Off mode: Indicates the MS is off. When off, all high voltages and the ion gauge are turned off. The sheath, auxiliary, and sweep gases remain on and return to their default settings (2 arbitrary units). When you remove the source housing or the spray insert, the MS automatically switches to Off mode. | |
Indicates the ion polarity mode (positive or negative). For more details, see Ion Polarity Modes. | |
Indicates the data type (profile or centroid). For more details, see Data Types. | |
Indicates whether the scan averaging feature is off or on. If scan averaging is turned on, enter a scan averaging value. | |
Indicates the valve position (1-2 or 1-6) of the optional divert/inject valve. The downward arrow opens the following options:
| |
Indicates whether the syringe pump is off or on. If the syringe pump is turned on, you can enter the syringe pump flow rate and syringe volume.
| |
Indicates the nitrogen collision cell gas pressure (in milliTorr). Range: 1–10; Default: 8 | |
A set of data acquisition control buttons to control the manual acquisition of sample data. For more details, see Data Acquisition Controls. | |
Readback Status Icon This is a read-only icon that changes to display the current MS instrument state. | |
Normal | Indicates that the MS instrument parameters are within tolerance. |
Initializing | Indicates that the system is initializing. |
Changing settings | Indicates that one or more settings are changing. |
Error | Indicates that an error has occurred. |
Ion source Off | Indicates that the ion source is off. |
Disconnected | Indicates that there is no communication between the MS and the data system. |
The following table describes the buttons and icons available at the bottom of the Tune application window.
Buttons | Description |
---|---|
Opens the Diagnostics pane. For more details, see Diagnostics Pane. | |
Opens the Plot Chromatogram dialog box. For more details, see Open the Plot Chromatogram Dialog Box. | |
Rotates the displayed plot. | |
Toggles between the three plot views (three plots, two plots, or a bottom plot). |