The following table lists the ion sources and interfaces that are compatible with the Stellar MS.
Compatible Ion Sources and Interfaces
Hardware | Recommended Flow Rate Range |
---|---|
OptaMax Plus ion source |
NOTE In APPI mode, you can use either the APCI spray insert or the H-ESI spray insert. |
Easy-Spray NG ion source | 10–1,000 nL/min |
Nanospray Flex NG ion source | 10–10,000 nL/min NOTE The flow rate range is dependent upon the type of column that is used. If used for a capillary flow, the flow rate range is 1–10 nL/min. |
VeriSpray PaperSpray ion source | N/A |
Interfaces | |
FAIMS Pro Duo interface | 100 nL/min – 1,000 μL/min NOTE When using a high flow rate of 1,000 μL/min, set the H-ESI spray insert or APCI spray insert to the high depth position. |
FAIMS Pro interface | 100–1,000 nL/min |