Item
Part Number
UHPLC/MS installation kit
HAZMAT-01-00140
or
UHPLCMSKIT
The kit contains the following items:
Water, UHPLC/MS-grade, 1 L (two provided)
W81
Methanol, UHPLC/MS-grade, 1 L (two provided)
A4581
Acetonitrile, UHPLC/MS-grade, 1 L
A9561
ChromaCare™ LC/MS instrument flush solution
T111101000