The following table shows the positions and includes the adjustment procedures for the spray insert within the API source.
Adjustment Control | Procedure |
---|---|
Front-to-back position | Adjust the Front-to-back Position Procedure
NOTE The OptaMax Plus ion source positions (0 mm, 5 mm, and 10 mm) are equivalent to the OptaMax NG ion source positions (1, 2, and 3). The OptaMax Plus ion source allows you to move the spray insert past the 0 mm mark for additional optimization of signal strength, if needed. |
Side-to-side position | Adjust the Side-to-side Position Procedure
NOTE This adjustment range is the same between the OptaMax Plus ion source and the OptaMax NG ion source. |
Depth position | Adjust the Depth Position Procedure
NOTE This adjustment range is the same between the OptaMax Plus ion source and the OptaMax NG ion source. |