Check the Auto-Ready components before you calibrate using Auto-Ready.
Procedure
- Verify that the Auto-Ready calibration solvent vial is seated in the vial receptacle and that the calibration vial locking lever is in the locked position.
- Verify that the PEEK fittings are properly tightened.
NOTE
If a fitting is loose, tighten it by hand (do not use tools). Over-tightening the PEEK fittings can cause leaks.
- Verify that the Auto-Ready sprayer is properly installed.
- Verify that the Auto-Ready calibration solvent vial level is full with the FlexMix solution.