Reinstall the API source interface after you have cleaned the internal components.
Prerequisites
  1. Put on a new pair of powder-free gloves.
Procedure
  1. Orient the API source interface so that the release latch is at the top.
  2. Carefully insert the API source interface into the vacuum manifold.
  3. Reinstall the API source housing.
  4. Start up the MS. See Restart the MS.