The following figure shows the installation of the L0 lens and MP00 RF rods onto the Exit lens.
Figure Alignment slot and thumbscrew locations on the MP00 RF rods
123456
No.DescriptionNo.Description
1
Exit lens
4
Slot for lead pin
2
MP00 RF rods
5
Recessed area where the thumbscew sits
3
L0 lens
6
Lead pin
Prerequisites
  1. Put on a new pair of powder-free gloves.
Procedure
  1. Align the slot for the lead pin on the MP00 RF rods with the lead pin on the Exit lens. Then carefully push the MP00 RF rods onto the Exit lens.
  2. Tighten the two thumbscrews so that they touch the MP00 RF rods.
  3. The thumbscrew should sit in the recessed area.
  4. Place the L0 lens on the MP00 RF rods.
  5. Insert the L0 lens removal tool (or plastic tweezer) into the L0 lens receptacles. Then rotate the L0 lens in the clockwise direction to secure it in place.