IMPORTANT
Do not clean the lenses with abrasives, acidic or caustic substances, or detergents that are not stated in this procedure.
Procedure
- Inspect the components for lint, particulates, and sample buildup or coatings using a magnification device.
- Use the 6,000 grit MICRO-MESH polishing swabs to clean the bore of the Exit lens, MP00 RF rods, and L0 lens.
- Sonicate the components in either a 50:50 solution of methanol-water or a 1% solution of Liquinox in water for 10 minutes.
- If a sonicator is not available, do the following:
- To clean the Exit lens, use a soft toothbrush with a 1% solution of Liquinox in water.
- Rinse the components thoroughly with water.
- Sonicate the components in water for 10 minutes.
- Sonicate the components in methanol for 10 minutes.
- Rinse the components with methanol.
- Dry the components thoroughly with nitrogen gas.
- Use a magnification device to inspect the components (bore of the orifices) for residual lint or particulates.
- If present, use plastic tweezers or a similar tool to remove them.