Remove API source interface to clean its internal components.
IMPORTANT
Risk of electric shock
Shut down the MS before you remove the API source interface to avoid the risk of an electric shock.
The following figure shows the API source interface in the vacuum manifold.
Figure API source interface
12
No.DescriptionNo.Description
1
Release latch on the API source interface
2
API source interface
Prerequisites
  1. Wear personal protective equipment (PPE) such as a lab coat, safety glasses, and chemical resistant gloves.
  2. Shut Down the MS.
  3. Remove the API Source.
Procedure
  1. Lift up on the release latch and firmly push it to unseal the API source interface from the vacuum manifold.
  2. Grasp the API source interface by the edges and carefully pull it out of the vacuum manifold.