The spray cone user maintenance is limited to cleaning its external surface. You do not need to remove the spray cone. Additionally, inspect and clean the ion transfer tube O-ring or replace it as needed.
The following figure shows the spray cone with the ion transfer tube O-ring, ion transfer tube, and ion sweep cone removed.
Figure Spray cone, O-ring, ion transfer tube, and ion sweep cone
1234
No.DescriptionNo.Description
1
Spray cone
3
Ion transfer tube
2
Ion transfer tube O-ring
4
Ion sweep cone
Prerequisites
  1. Wear personal protective equipment (PPE) such as a lab coat, safety glasses, and chemical resistant gloves.
  2. Remove the Ion Sweep Cone
  3. Remove the Ion Transfer Tube
Procedure
  1. Soak a lint-free tissue or chamois-tipped swab in a 50:50 solution of methanol-water, and then clean the exterior surface of the spray cone.
  2. Using a magnification device, inspect the spray cone for residual lint or particulates.
  3. If present, use plastic tweezers or a similar tool to remove them.
  4. Remove and inspect the ion transfer tube O-ring located at the spray cone entrance. Clean it with methanol or replace it if necessary.
  5. IMPORTANT
    Be careful not to drop or damage the O-ring during the removal or installation.