CAUTION
Avoid exposure to potentially harmful materials
By law, producers and suppliers of chemical compounds are required to provide their customers with the most current health and safety information in the form of Material Safety Data Sheets (MSDSs) or Safety Data Sheets (SDSs). The MSDSs and SDSs must be freely available to lab personnel to examine at any time. These data sheets describe the chemicals and summarize information on the hazard and toxicity of specific chemical compounds. They also provide information on the proper handling of reagents, first aid for accidental exposure, and procedures to remedy spills or leaks.
CAUTION
Risk of eye injury
Read the MSDS or SDS for each chemical you use. Store and handle all chemicals in accordance with standard safety procedures. Always wear protective gloves and safety glasses when you use solvents or corrosives. Also, contain waste streams, use proper ventilation, and dispose of all laboratory reagents according to the directions in the MSDS or SDS.
The MS requires very few tools to perform routine maintenance. You can remove and disassemble many of the components by hand. The following table lists the necessary chemicals, tools, and equipment for maintaining the instrument.
IMPORTANT
Always use UHPLC/MS-grade methanol and water.
For a wide variety of solvents and consumables, go to www.fishersci.com.
Chemicals, Tools, and Equipment
Description | Part Number |
---|---|
Chemicals | |
Liquinox™, Citranox™, or similar detergent | – |
Nitrogen gas, clean and dry | – |
Methanol, UHPLC/MS-grade, 1 L | |
Water, UHPLC/MS-grade, 1 L | |
Tools | |
Ion transfer tube removal tool (provided) | 70005-20972 |
L0 lens removal tool (provided) | 70005-20900 |
Screwdriver, Phillips #2 | – |
Soft toothbrush or similar tool | – |
Plastic tweezer or similar tool | – |
Equipment | |
Gloves, nitrile powder-free | 19-120-29471) |
Chamois-tipped swabs | 00725-01-00028 |
Industrial tissues, lint-free | – |
MICRO-MESH™ polishing swabs, | 00725-01-00027 |
Beaker or graduated cylinder (for use with methanol) | – |
Magnifier | – |
Sonicator | – |